Learn More
Qorpak™ ValuLine F217 and PTFE Lined Polypropylene Caps Available on GSA/VA Contract for Federal Government customers only.
Help to keep bottle or jar contents free of odor and taste interference
Manufacturer:  Qorpak™ 239075
 View more versions of this product
Catalog No. 02-991-377
  Check Availability
Add to cart
Description
Description
  • Made of white polypropylene with PTFE facing
  • F217 liner backing consisting of low density foam core tightly sandwiched between two layers of LDPE
  • Resilient with strong resistance to chemicals and moisture
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
F217 and PTFE
8500 Cs.
Wide mouth bottle
Ribbed
White
Bulk
22-400