Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White HDPE Cyclinder Bottles, With White Polypropylene SturdeeSeal™ PE Foam Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Offers a degree of light protection

Manufacturer:  Qorpak™ PLC05766

 View more versions of this product

Catalog No. 02-991-285


  Check Availability
Add to cart

Description

Description

  • Impact resistant
  • Provides moisture barrier
  • Narrow mouth: Makes handling of liquid samples easy
  • Shoulder style allows to pour small quantities without spilling
  • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

6 oz., 180mL
6 oz.
SturdeeSeal™ Polyethylene Foam
1.77 in.
HDPE
White
45mm
24-410
180mL
Polypropylene
White
5.94 in.
300/Cs.
24-410
151mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit