Qorpak™ White Metal Plastisol Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Tolerates wide ranges of temperature

Manufacturer: Qorpak™ CAP00450

 View more versions of this product

Catalog No. 02-991-062

  Check Availability
Add to Cart

    Specifications

    Closure Liner Plastisol
    Color White
    For Use With Wide mouth bottle
    Neck Finish 53-400
    No. per Case 1900 Cs.
    No. per Pack 1900
    Packaging Bulk

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit