Learn More
Qorpak™ White Metal Plastisol Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Tolerates wide ranges of temperature
Manufacturer:  Qorpak™ CAP00459
 View more versions of this product
Catalog No. 02-991-032
  Check Availability
Add to cart
Description
Description
  • Sturdy and unbreakable
  • Impact resistant
  • Made of steel coated with anti-corrosive tinplate
  • Permit hot fill operations to produce vacuum seal
  • Provide resistance to mild acids as in food products
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Plastisol
1700 Cs.
Bulk
63-400
White
1700
Wide mouth bottle