Learn More
Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Free of odor and taste problems
Manufacturer:  Qorpak™ CAP06648
 View more versions of this product
Catalog No. 02-993-392
  Check Availability
Add to cart
Description
Description
  • Made of steel coated with anti-corrosive tinplate
  • Impact resistant
  • Tolerate wide ranges of temperature
  • Feature standard 400 GPI thread finish
  • Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Pulp/Aluminum Foil
576 Cs.
Bagged
33-400
White
12/Pk.
Wide mouth bottle