Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps

Catalog Number 02991110
Click to view available options
Neck Finish:
24-400
28-400
33-400
38-400
43-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
1400 Cs.
144 Cs.
2000 Cs.
288 Cs.
3000 Cs.
3400 Cs.
5400 Cs.
576 Cs.
800 Cs.
Packaging:
Bagged
Bulk

Free of odor and taste problems

  • Made of steel coated with anti-corrosive tinplate
  • Impact resistant
  • Tolerate wide ranges of temperature
  • Feature standard 400 GPI thread finish
  • Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils

Closure Liner Pulp/Aluminum Foil
For Use With (Equipment) Wide mouth bottle
Color White
Material Metal
No. per Case 2000 Cs.
Packaging Bulk
Neck Finish 63-400
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit