Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Free of odor and taste problems

Manufacturer: Qorpak™ CAP00443

 View more versions of this product

Catalog No. 02-991-130

  Check Availability
Add to Cart

    Specifications

    Closure Liner Pulp/Aluminum Foil
    Color White
    For Use With Wide mouth bottle
    Neck Finish 43-400
    No. per Case 3100 Cs.
    Packaging Bulk

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit