Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ White Metal Pulp/Polyethylene Lined Caps
Click to view available options
Neck Finish:
100-400
33-400
38-400
43-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
1400 Cs.
144 Cs.
288 Cs.
500 Cs.
576 Cs.
Packaging:
Bagged
Bulk
Description
- Resistant to fracture from impact
- Offered in 400 GPI thread finish
- For general-purpose
- Ideal for powders, non-acidic food products and consumer products
Specifications
Closure Liner | Pulp/Polyethylene |
For Use With (Equipment) | Wide mouth bottle |
Color | White |
Material | Metal |
No. per Case | 144 Cs. |
No. per Pack | 12/Pk. |
Packaging | Bagged |
Neck Finish | 89-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction