Learn More
Qorpak™ White Metal Pulp/Polyethylene Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Exhibit tolerance to widest range of temperatures
Manufacturer:  Qorpak™ CAP00440
 View more versions of this product
Catalog No. 02-991-190
  Check Availability
Add to cart
Description
Description
  • Resistant to fracture from impact
  • Offered in 400 GPI thread finish
  • For general-purpose
  • Ideal for powders, non-acidic food products and consumer products
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Pulp/Polyethylene
3800 Cs.
Wide mouth bottle
White
Bulk
38-400