Learn More
Qorpak™ White Polypropylene Unlined Caps Available on GSA/VA Contract for Federal Government customers only.
Suitable for various kinds of dry products
Manufacturer:  Qorpak™ CAP06638
 View more versions of this product
Catalog No. 02-992-901
  Check Availability
Add to cart
Description
Description
  • High impact strength
  • Pliable
  • Autoclavable
  • Available in ribbed style
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
White
Bagged
58-400
288 Cs.
Wide mouth bottle
Ribbed