Learn More
Qorpak™ White Polypropylene Unlined Caps
Suitable for various kinds of dry products
Manufacturer:  Qorpak™ CAP06639
 View more versions of this product
Catalog No. 02-992-983
Add to cart
Description
Description
  • High impact strength
  • Pliable
  • Autoclavable
  • Available in ribbed style
Specifications
Specifications
White
288 Cs.
Wide mouth bottle
Ribbed
Polypropylene
Bagged
63-400