Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Unlined Polypropylene Caps
Click to view available options
Neck Finish:
120-400
33-400
33-430
38-400
43-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
1100 Caps/Cs.
1300 Caps/Cs.
144 Caps/Cs.
1700 Caps/Cs.
1800 Caps/Cs.
2200 Caps/Cs.
242 Caps/Cs.
288 Caps/Cs.
2900 Caps/Cs.
4000 Caps/Cs.
456 Caps/Cs.
576 Caps/Cs.
72 Caps/Cs.
760 Caps/Cs.
900 Caps/Cs.
Description
- Autoclavable
- Well-suited for many dry products
- Add a PTFE disc liner to the unlined polypropylene cap for better leak resistance when autoclaving
Specifications
For Use With (Equipment) | Wide Mouth Bottle |
Closure Type | Ribbed |
Color | White |
Material | Polypropylene |
No. per Case | 1100 Caps/Cs. |
Packaging | Bulk |
Neck Finish | 58-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction