Learn More
MP Biomedicals™ qPCR& Go Probe No-ROX Kit
Fast and sensitive qPCR Probe Master Mix. For all other platforms, e.g. Roche, Bio-Rad, Qiagen, etc.
Manufacturer:  MP Biomedicals™ 11EBI06200
 View more versions of this product
Catalog No. MP1EBI06200
Add to cart
Description
Description
  • The qPCR& Go Probe No-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
  • The kit has been formulated for use with probe-detection technology, including TaqMan™, Scorpions™ and molecular beacon probes.
  • A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCR& Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
  • qPCR& Go Probe is provided as a 2x mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
Specifications
Specifications
2000 reactions
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.