Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ QPCT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543215
Description
Peptide sequence: SRHLAAKMAS TPHPPGARGT SQLHGMDLLV LLDLIGAPNP TFPNFFPNSA Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 100%; Sheep: 86%.
GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
Specifications
QPCT | |
Polyclonal | |
Unconjugated | |
Qpct | |
5730422A13Rik; EC; GCT; Glutaminyl cyclase; glutaminyl-peptide cyclotransferase; glutaminyl-peptide cyclotransferase (glutaminyl cyclase); glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC; Qpct; RGD1562284; similar to Glutaminyl-peptide cyclotransferase precursor (QC); sQC | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25797 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q16769 | |
Qpct | |
Synthetic peptide directed towards the middle region of human QPCT. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction