Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QPCTL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548955

Catalog No. PA548955


Add to cart

Description

Description

Predicted to react with monkey based on sequence homology.

Responsible for the biosynthesis of pyroglutamyl peptides.
Specifications

Specifications

QPCTL
Polyclonal
PBS with 0.09% sodium azide
Q9NXS2
Glutaminyl-peptide cyclotransferase-like protein, Golgi-resident glutaminyl-peptide cyclotransferase, isoQC, gQC, QPCTL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54814
Western Blot
Unconjugated
QPCTL
Liquid
QPCTL
KLH conjugated synthetic peptide between 271-297 amino acids from the C-terminal region of human QPCTL.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit