Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QPRT Mouse anti-Human, Clone: OTI4E6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525199

Catalog No. PIMA525199


Add to cart

Description

Description

This protein is a key enzyme in catabolism of quinolinate. an intermediate in the tryptophan-nicotinamide adenine dinucleotide pathway. Quinolinate acts as a most potent endogenous exitotoxin to neurons. Elevation of quinolinate levels in the brain has be.
Specifications

Specifications

QPRT
Monoclonal
0.83 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q15274
Nicotinate-nucleotide pyrophosphorylase [carboxylating], Quinolinate phosphoribosyltransferase [decarboxylating], QAPRTase, QPRTase, QPRT
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23475
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI4E6
Unconjugated
QPRT
Liquid
QPRT
Full length human recombit protein of QPRT produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit