Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QRP™ Qualatex™ 2C Low-Powder Latex Finger Cots
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- General protection against dyes, chemical splashes, and solvents in addition to skin salts, flakes, and oils
- Sheer film ensures maximum dexterity
- For industrial use only
Specifications
Applications | Industrial |
Color | White |
Disposable | Yes |
Exterior Finish | Partially Textured |
Hand | Ambidextrous |
Material | Latex |
Powdered | Powdered |
Product Type | Finger Cot |
Size | Small |
Thickness (English) | 3.5 mil |
Show More |