Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QRP™ BQF09 Qualatrile™ 5 mil Blue Nitrile Powder Free Gloves

Catalog Number 19048740B
Click to view available options
Size:
X-Small
Small
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Powder free, 5 mil nitrile gloves are naturally anti-static and offer 3x more puncture resistance than vinyl or natural rubber latex gloves.

  • 5 mil thickness
  • 3x puncture resistance of vinyl or natural rubber latex gloves
  • Naturally anti-static, surface resistance 10(9) ohms
  • Geogrip 360 sm fully textured fingers and palms
  • Meets ASTM D6319
Certifications/Compliance ASTM D6319
Applications Laboratory, Industrial
Color Blue
Exterior Finish Fully Textured
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Size Small
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
Model BQF09S
Series Qualatrile™ Blue 5
Length (English) 9 in.
Length (Metric) 22.86 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit