Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QRP™ BQP09 Qualatrile™ Low Powder Blue Nitrile Gloves

Catalog Number p-4717776
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Low powder, 5 mil thick nitrile gloves are naturally anti-static and fully textured for a variety of general purpose applications.

  • 5 mil thickness
  • 3x puncture resistance of vinyl or natural rubber latex gloves
  • Naturally anti-static, surface resistance 10(9) ohms
  • Geogrip 360 sm fully textured fingers and palms
  • Meets ASTM D6319
Certifications/Compliance ASTM D6319
Applications Laboratory, Industrial
Color Blue
Exterior Finish Fully Textured
Hand Ambidextrous
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Powdered Powdered
Size Medium
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
Model BQP09M
Series Qualatrile™ Blue 5
Length (English) 9 in.
Length (Metric) 22.86 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit