Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QRP™ Qualatrile™ Blue Nitrile 9 in Powder-Free Gloves

Manufacturer:  QRP™ BQF09XS

 View more versions of this product

Catalog No. 19-048-740A



Add to cart

Specifications

Specifications

Safety
Beaded/Rolled
Nitrile, Rubber Latex, Vinyl
4 mil
Blue
22.86 cm
Qualatrile blue Nitrile gloves
Ambidextrous
X-Small
Powder-free
9 in.
ASTM D6319, RoHS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit