QRP™ Qualatrile™ Blue Nitrile 9 in Powder-Free Gloves

Manufacturer: QRP™ 8BQF09M

 View more versions of this product

Catalog No. 19-048-746C

  Check Availability
Add to Cart

    Specifications

    Certifications ASTM D6319, RoHS
    Glove Cuff Style Beaded
    Hand Ambidextrous
    Length (English) 9 in.
    Length (Metric) 22.86cm
    Material Nitrile, Natural Rubber Latex, Vinyl
    Powdered No
    Size Medium
    Thickness 8mil
    Color Blue
    Classification Qualatrile blue Nitrile gloves

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit