Learn More
QRP™ Static Dissipative Latex Finger Cots
For Class I static-sensitive devices
Manufacturer:  QRP™ 8JM
Catalog No. 19-076196
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
3mil; measure 2 3/4 to 3 in. (7 to 7.6cm) long. In small, medium, large, and x-large sizes. Packaged 1440 cots per pack.
  • Rolled cots have average nongenerative surface resistivity of 107 ohms/sq. in.
  • Dissipate charge in under 0.25 second
  • Suggested for Class I (0-1999V) static-sensitive devices
  • Also appropriate for Class II devices
  • Made of washed, powder-free latex with a proprietary dissipative compound
  • Sheer, 3mil; 2 3/4 to 3 in. long (7 to 7.6cm)
Order Info
Double packed in ESD-safe inner and outer wrap; immobilized in nitrogen-flushed TrayPak™ to prevent charge generation. Packaged 720 cots per pack.
Compliance
Comply with MIL-B-81705, DOD-HDBK-263, and MIL-STD-1686D. For ISO Class 5 (formerly Class 100) cleanrooms.
Specifications
Specifications
Safety