Learn More
Qsonica Sonicator Q500 Available on GSA/VA Contract for Federal Government customers only.
Popular applications include nanoparticle dispersion, creating emulsions, cell lysis and homogenization
Manufacturer:  Qsonica Q500220
 View more versions of this product
Catalog No. 15-338-415
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
Ideal for nano particle dispersion, creating emulsions, cell lysis and homogenization
21.59cm
Generator, converter, power cable, converter cable, wrench set, probe
38.73cm
220V
8 in.
15.25 x 8 x 8.5 in. (38.73 x 20.32 x 21.59cm)
8.5 in.
50/60Hz
15.25 in.
10 hrs.
500w
20.32cm
220V 50/60Hz