Learn More
Qsonica Ultrasonic Homogenizer Converters Available on GSA/VA Contract for Federal Government customers only.
Qsonica Ultrasonic Homogenizer Converter works with either the Microson XL2000 or the Sonicator 3000 Ultrasonic Homogenizers to transform electrical signals into mechanical vibrations.
Manufacturer:  Qsonica CL5
 View more versions of this product
Catalog No. 15-338-261
  Check Availability
Add to cart
Specifications
Description
  • For use with Microson? Model XL2000 or Sonicator? 3000 Ultrasonic Homgenizers
  • Transforms electrical signal into mechanical vibrations
Description & Specifications
Specifications
3000 and 4000 Sonicator™ ultrasonic homogenizer
Converter
Ideal for transform electrical signal into mechanical vibration