Learn More
QSOX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA590300
Description
Immunogen sequence: VKKLLEMLQE WLASLPLDRI PYNAVLDLVN NKMRISGIFL TNHIKWVGCQ GSRSELRGYP CSLWKLFHTL TVEASTHPDA LVGTGFEDDP QAVLQTMRRY VHTFFGCKEC GEHFEEMAKE SMDSVKTPDQ AILWLWKKHN MVNGRLAGHL SEDPRFPKLQ WPTPDLCPAC HEEIKGLASW DEGHVLTFLK QHYGRDNLLD TYSADQGDSS EGGTLARGEE EEKRLTPPEV SHGDRDTQSV RPPGALGPRP ALPESLHHSL DGKLQSLDGP GAHKEVGGAA P; Positive Samples: Mouse pancreas, Mouse testis; Cellular Location: Cell membrane, Membrane, Nucleus membrane, Single-pass membrane protein
QSOX2 is a member of the sulfhydryl oxidase/quiescin-6 (Q6) family (QSOX1) that regulates the sensitization of neuroblastoma cells for IFN-gamma (IFNG)-induced cell death.Specifications
QSOX2 | |
Polyclonal | |
Unconjugated | |
Qsox2 | |
Qsox2 | |
Recombit fusion protein containing a sequence corresponding to amino acids 370-650 of human QSOX2 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
2.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TMX7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227638 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.