Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human QTRTD1 His Protein
SDP

Highly purified. Generating reliable and reproducible results.

$494.50 - $1472.00

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffer saline (pH 7.4) containing 30% glycerol, 2mM DTT, 1mM EDTA, 0.1mM PMSF
Gene ID (Entrez) 79691
Molecular Weight (g/mol) TMW: 49.1kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25187805
SDP
View Documents
Novus Biologicals™
NBP2518780.05MG
0.05mg
Each for $494.50
Add to Cart
 
NBP25187825
SDP
View Documents
Novus Biologicals™
NBP2518780.25MG
0.25mg
Each for $1,472.00
Add to Cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-415 of Human QTRTD1 The Recombinant Human QTRTD1 Protein is derived from E. coli. The Recombinant Human QTRTD1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25mg/mL
Phosphate buffer saline (pH 7.4) containing 30% glycerol, 2mM DTT, 1mM EDTA, 0.1mM PMSF
TMW: 49.1kDa
EC 2.4.2.29, FLJ12960, queuine tRNA-ribosyltransferase domain containing 1, Queuine tRNA-ribosyltransferase domain-containing protein 1, queuine tRNA-ribosyltransferase subunit QTRTD1
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
SDS-PAGE
79691
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
QTRTD1
Recombinant
SDS
Documents

Documents

Product Certifications

For research use only and is not approved for use in humans or in clinical diagnosis

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit