Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuadraPure™ AK, 50-90 mesh, metal scavenger, Thermo Scientific™

Scavenger for Hydrazines and Primary Amines

Specifications

Chemical Name or Material QuadraPure™ AK metal scavenger
MDL Number MFCD03792524
Synonym 2,4-Butanedione resin, Activated ketone, polymer-bound
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA4628314
View Documents
Thermo Scientific Chemicals
4628314
25g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
AA4628306
View Documents
Thermo Scientific Chemicals
4628306
5g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

QuadraPure™ AK metal scavenger
MFCD03792524
2,4-Butanedione resin, Activated ketone, polymer-bound
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit