Learn More
Alfa Aesar™ QuadraPure™ AK, 50-90 mesh, metal scavenger
Scavenger for Hydrazines and Primary Amines
Manufacturer:  Alfa Aesar™ 4628306
Quantity 5g
 View more versions of this product
Catalog No. AA4628306
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
MFCD03792524
2,4-Butanedione resin, Activated ketone, polymer-bound
QuadraPure™ AK metal scavenger
Specifications
Beads
2,4-Butanedione resin, Activated ketone, polymer-bound
White to beige
QuadraPure™ AK metal scavenger
MFCD03792524
50 to 90 Mesh
5g