Alfa Aesar™ QuadraPure™ BDZ, 400-750 micron, metal scavenger

Metal Scavenger: Rh, Co, Pd, Ni

Manufacturer: Alfa Aesar™ 4600822

Quantity 100g

 View more versions of this product

Catalog No. AA4600822

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD09039244
    Synonym QuadraPure™ Bis(propyl-1H-imidazole)
    Chemical Name or Material QuadraPure™ BDZ metal scavenger

    Specifications

    Synonym QuadraPure™ Bis(propyl-1H-imidazole)
    Physical Form Beads
    MDL Number MFCD09039244
    Particle Size 400 to 750 Micron
    Color White to beige
    Quantity 100g
    Chemical Name or Material QuadraPure™ BDZ metal scavenger

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit