Alfa Aesar™ QuadraPure™ BZA, 400-1100 micron, metal scavenger

Metal Scavenger: Rh, Pd, Cu, Co, Ni

Manufacturer: Alfa Aesar™ 4598922

Quantity 100g

 View more versions of this product

Catalog No. AA4598922

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD08561140
    UN Number UN3082
    Synonym QuadraPure™ Benzylamine
    Chemical Name or Material QuadraPure™ BZA metal scavenger

    Specifications

    Synonym QuadraPure™ Benzylamine
    Physical Form Beads
    UN Number UN3082
    Particle Size 400 to 1100 Micron
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Quantity 100g
    MDL Number MFCD08561140
    Color White to beige
    Chemical Name or Material QuadraPure™ BZA metal scavenger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit