Alfa Aesar™ QuadraPure™ C, 0.3-0.8mm, metal scavenger

Manufacturer: Alfa Aesar™ 4629730

Quantity 250g

 View more versions of this product

Catalog No. AA4629730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-44-0
    MDL Number MFCD09753896
    Chemical Name or Material QuadraPure™ C, 0.3 to 0.8mm, metal scavenger

    Specifications

    Physical Form Beads
    MDL Number MFCD09753896
    CAS 7440-44-0
    Color Black
    Quantity 250g
    Chemical Name or Material QuadraPure™ C, 0.3 to 0.8mm, metal scavenger

    Description

    • High-capacity, carbon-based adsorbent in the form of spherical particles used for scavenging of trace organic impurities
    • Free-flowing and free of dust, QuadraPure™ C is easily removed from reaction mixtures by filtration

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit