Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ QuadraPure™ C, 0.3-0.8mm, metal scavenger

Manufacturer:  Alfa Aesar™ 4629718

Quantity 50g

 View more versions of this product

Catalog No. AA4629718


  Check Availability
Add to cart

Specifications

Description

  • High-capacity, carbon-based adsorbent in the form of spherical particles used for scavenging of trace organic impurities
  • Free-flowing and free of dust, QuadraPure™ C is easily removed from reaction mixtures by filtration
Description & Specifications

Chemical Identifiers

7440-44-0
MFCD09753896
QuadraPure™ C, 0.3 to 0.8mm, metal scavenger

Specifications

7440-44-0
MFCD09753896
50g
Beads
Black
QuadraPure™ C, 0.3 to 0.8mm, metal scavenger
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit