Alfa Aesar™ QuadraPure™ DET, 450-650 micron, metal scavenger

Metal Scavenger: Pd in acidic media and with phosphines, Co, Fe, Ni, Rh

Manufacturer: Alfa Aesar™ 46083-14

Quantity 25g

 View more versions of this product

Catalog No. AA4608314

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD08705362
    Synonym QuadraPure™ Bis(ethyl mercaptan)
    Chemical Name or Material QuadraPure™ DET metal scavenger

    Specifications

    Synonym QuadraPure™ Bis(ethyl mercaptan)
    Physical Form Beads
    MDL Number MFCD08705362
    Particle Size 450 to 650 Micron
    Color White to beige
    Quantity 25g
    Chemical Name or Material QuadraPure™ DET metal scavenger

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit