Learn More
Alfa Aesar™ QuadraPure™ DET, 450-650 micron, metal scavenger
Metal Scavenger: Pd in acidic media and with phosphines, Co, Fe, Ni, Rh
Manufacturer:  Alfa Aesar™ 4608306
Quantity 5g
 View more versions of this product
Catalog No. AA4608306
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
MFCD08705362
QuadraPure™ Bis(ethyl mercaptan)
QuadraPure™ DET metal scavenger
Specifications
Beads
QuadraPure™ Bis(ethyl mercaptan)
White to beige
QuadraPure™ DET metal scavenger
MFCD08705362
450 to 650 Micron
5g