Alfa Aesar™ QuadraPure™ DMA, 400-800 micron, metal scavenger

Metal scavenger: Ag, Au, Cu, Fe, Ir, Ni, Pd, Pt, Rh

Manufacturer: Alfa Aesar™ 4632622

Quantity 100g

 View more versions of this product

Catalog No. AA4632622

  Check Availability
Add to Cart

    Chemical Identifiers

    Synonym QuadraPure™ Dimethylamine
    Chemical Name or Material QuadraPure™ DMA metal scavenger

    Specifications

    Synonym QuadraPure™ Dimethylamine
    TSCA TSCA
    Physical Form Beads
    Quantity 100g
    Particle Size 400 to 800 Micron
    Color Off-white
    Chemical Name or Material QuadraPure™ DMA metal scavenger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit