Alfa Aesar™ QuadraPure™ EDA, 500-800 micron, metal scavenger

Metal Scavenger: Pd in basic media and with phosphines, Co, Ni, Rh

Manufacturer: Alfa Aesar™ 4604414

Quantity 25g

 View more versions of this product

Catalog No. AA4604414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD09039245
    Synonym QuadraPure™ Bis(ethylamine)
    Chemical Name or Material QuadraPure™ EDA metal scavenger

    Specifications

    Synonym QuadraPure™ Bis(ethylamine)
    MDL Number MFCD09039245
    Color White to beige
    Physical Form Beads
    Particle Size 500 to 800 Micron
    Quantity 25g
    Chemical Name or Material QuadraPure™ EDA metal scavenger

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit