Learn More
Alfa Aesar™ QuadraPure™ EDA, 500-800 micron, metal scavenger
Metal Scavenger: Pd in basic media and with phosphines, Co, Ni, Rh
Manufacturer:  Alfa Aesar™ 4604406
Quantity 5g
 View more versions of this product
Catalog No. AA4604406
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
MFCD09039245
QuadraPure™ Bis(ethylamine)
QuadraPure™ EDA metal scavenger
Specifications
Beads
QuadraPure™ Bis(ethylamine)
White to beige
QuadraPure™ EDA metal scavenger
MFCD09039245
500 to 800 Micron
5g