Alfa Aesar™ QuadraPure™ IDA, 350-750 micron, metal scavenger

Metal Scavenger: Fe, Al, Ga, In, Cu, V, Pb, Ni, Zn, Cd, Be, Mn, Co, Sr, Ba

Manufacturer: Alfa Aesar™ 4595206

Quantity 5g

 View more versions of this product

Catalog No. AA4595206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 135620-93-8
    Formula Weight 0.73
    MDL Number MFCD07785598
    UN Number UN3175
    Synonym QuadraPure™ Iminodiacetate
    Chemical Name or Material QuadraPure™ IDA metal scavenger

    Specifications

    Formula Weight 0.73
    Synonym QuadraPure™ Iminodiacetate
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    CAS 135620-93-8
    UN Number UN3175
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    Particle Size 350 to 750 Micron
    Color White to pale cream
    Quantity 5g
    Physical Form Beads
    MDL Number MFCD07785598
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: SOLIDS CONTAINING FLAMMABLE LIQUID, N.O.S.
    Chemical Name or Material QuadraPure™ IDA metal scavenger

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit