Learn More
Alfa Aesar™ QuadraPure™ IDA, 350-750 micron, metal scavenger
Metal Scavenger: Fe, Al, Ga, In, Cu, V, Pb, Ni, Zn, Cd, Be, Mn, Co, Sr, Ba
Manufacturer:  Alfa Aesar™ 4595214
Quantity 25g
 View more versions of this product
Catalog No. AA4595214
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
135620-93-8
MFCD07785598
QuadraPure™ Iminodiacetate
0.73
UN3175
QuadraPure™ IDA metal scavenger
Specifications
135620-93-8
0.73
UN3175
350 to 750 Micron
25g
Beads
MFCD07785598
QuadraPure™ Iminodiacetate
White to pale cream
QuadraPure™ IDA metal scavenger
Safety and Handling
SDS & Safety Documents
GHS H Statement
H228
Flammable solid.
GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: SOLIDS CONTAINING FLAMMABLE LIQUID, N.O.S.