Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuadraPure™ IMDAZ, 100-400 micron, metal scavenger, Thermo Scientific™

Metal Scavenger: Pd, Ru, Os, Co, Ni, Rh, V, Fe, Cu, Sn

Specifications

Chemical Name or Material QuadraPure™ IMDAZ metal scavenger
MDL Number MFCD07785599
UN Number UN3082
Synonym QuadraPure™ Imidazol-1-yl propylaminobut-2-enoate ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA4613306
View Documents
Thermo Scientific Chemicals
4613306
5g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
AA4613314
View Documents
Thermo Scientific Chemicals
4613314
25g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

QuadraPure™ IMDAZ metal scavenger
MFCD07785599
UN3082
QuadraPure™ Imidazol-1-yl propylaminobut-2-enoate ester
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit