Learn More
Alfa Aesar™ QuadraPure™ SA, 400-850 micron, metal scavenger
Manufacturer:  Alfa Aesar™ 4639214
Quantity 25g
 View more versions of this product
Catalog No. AA4639214
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
UN3175
QuadraPure™ Sulfonic acid
QuadraPure™ SA metal scavenger
Specifications
Beads
QuadraPure™ Sulfonic acid
Off-white to pale brown
QuadraPure™ SA metal scavenger
UN3175
400 to 850 Micron
25g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H228-H318
Flammable solid.
Causes serious eye damage.
GHS P Statement
P210-P280-P240-P241-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: SOLIDS CONTAINING FLAMMABLE LIQUID, N.O.S.