Alfa Aesar™ QuadraPure™ SA, 400-850 micron, metal scavenger

Manufacturer: Alfa Aesar™ 4639222

Quantity 100g

 View more versions of this product

Catalog No. AA4639222

  Check Availability
Add to Cart

    Chemical Identifiers

    UN Number UN3175
    Synonym QuadraPure™ Sulfonic acid
    Chemical Name or Material QuadraPure™ SA metal scavenger

    Specifications

    Synonym QuadraPure™ Sulfonic acid
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    UN Number UN3175
    Physical Form Beads
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H318
    Flammable solid.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: SOLIDS CONTAINING FLAMMABLE LIQUID, N.O.S.
    Color Off-white to pale brown
    Particle Size 400 to 850 Micron
    Quantity 100g
    Chemical Name or Material QuadraPure™ SA metal scavenger

    GHS H Statement
    H228-H318
    Flammable solid.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P240-P241-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit