Alfa Aesar™ QuadraSil™ AP, 20-100 micron, metal scavenger

Metal scavenger: Co, Cu, Cd, Fe, Ni, Pd, Rh, Ru, Pb, Hg, Zn

Manufacturer: Alfa Aesar™ 4630314

Quantity 25g

 View more versions of this product

Catalog No. AA4630314

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD09753897
    Synonym QuadraSil™ Aminopropyl
    Chemical Name or Material QuadraSil™ AP metal scavenger

    Specifications

    Synonym QuadraSil™ Aminopropyl
    Physical Form Powder
    MDL Number MFCD09753897
    Particle Size 20 to 100 Micron
    Color White
    Quantity 25g
    Chemical Name or Material QuadraSil™ AP metal scavenger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit