Learn More
Alfa Aesar™ QuadraSil™ AP, 20-100 micron, metal scavenger
Metal scavenger: Co, Cu, Cd, Fe, Ni, Pd, Rh, Ru, Pb, Hg, Zn
Manufacturer:  Alfa Aesar™ 4630322
Quantity 100g
 View more versions of this product
Catalog No. AA4630322
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD09753897
QuadraSil™ Aminopropyl
QuadraSil™ AP metal scavenger
Specifications
Powder
QuadraSil™ Aminopropyl
White
QuadraSil™ AP metal scavenger
MFCD09753897
20 to 100 Micron
100g