Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Alfa Aesar™ QuadraSil™ AP, 20-100 micron, metal scavenger
Metal scavenger: Co, Cu, Cd, Fe, Ni, Pd, Rh, Ru, Pb, Hg, Zn
Manufacturer: Alfa Aesar™ 4630306
Quantity 5g
 View more versions of this product
Catalog No. AA4630306
  N/A
Add to Cart
Related Products
Product Content Feedback