Alfa Aesar™ QuadraSil™ MP, 20-100 micron, metal scavenger

Metal scavenger: Pd (with or without phosphines), Rh, Cu, Ru, Pt, Pb, Ag, Hg

Manufacturer: Alfa Aesar™ 4625906

Quantity 5g

 View more versions of this product

Catalog No. AA4625906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1225327-73-0
    MDL Number MFCD09753898
    Synonym QuadraSil™ Mercaptopropyl
    Chemical Name or Material QuadraSil™ MP metal scavenger

    Specifications

    Synonym QuadraSil™ Mercaptopropyl
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder or Beads
    Health Hazard 1 Warning
    MDL Number MFCD09753898
    CAS 1225327-73-0
    Health Hazard 2 GHS H Statement
    H302-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Particle Size 20 to 100 Micron
    Color White to cream
    Quantity 5g
    Chemical Name or Material QuadraSil™ MP metal scavenger

    GHS H Statement
    H302-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit