Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ QuadraSil™ MTU, 20-100 micron, metal scavenger

Metal scavenger: Co, Cu, Fe, Pb, Pd, Rh, Ru, Ag, Au, Pt, Hg

Specifications

Chemical Name or Material QuadraSil™ MTU metal scavenger
MDL Number MFCD09753899
Synonym QuadraSil™ Methylthiourea
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA4618806 View Documents Alfa Aesar™
4618806
5g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
AA4618814 View Documents Alfa Aesar™
4618814
25g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
AA4618822 View Documents Alfa Aesar™
4618822
100g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications

Specifications

QuadraSil™ MTU metal scavenger
MFCD09753899
QuadraSil™ Methylthiourea
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit