Alfa Aesar™ QuadraSil™ TA, 20-100 micron, metal scavenger

Metal scavenger: Au, Cd, Co, Cu, Fe, Pd, Pt, Rh, Ru, V, Zn, Pb

Manufacturer: Alfa Aesar™ 4621814

Quantity 25g

 View more versions of this product

Catalog No. AA4621814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1187636-37-8
    MDL Number MFCD09753900
    Synonym QuadraSil™ Triamine
    Chemical Name or Material QuadraSil™ TA metal scavenger

    Specifications

    Synonym QuadraSil™ Triamine
    MDL Number MFCD09753900
    CAS 1187636-37-8
    Particle Size 20 to 100 Micron
    Quantity 25g
    Physical Form Powder or Beads
    Color White to beige
    Chemical Name or Material QuadraSil™ TA metal scavenger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit