Learn More
Alfa Aesar™ QuadraSil™ TA, 20-100 micron, metal scavenger
Metal scavenger: Au, Cd, Co, Cu, Fe, Pd, Pt, Rh, Ru, V, Zn, Pb
Manufacturer:  Alfa Aesar™ 4621822
Quantity 100g
 View more versions of this product
Catalog No. AA4621822
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
1187636-37-8
MFCD09753900
QuadraSil™ Triamine
QuadraSil™ TA metal scavenger
Specifications
1187636-37-8
MFCD09753900
20 to 100 Micron
100g
Powder or Beads
QuadraSil™ Triamine
White to beige
QuadraSil™ TA metal scavenger