Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Alfa Aesar™ QuadraSil™ TA, 20-100 micron, metal scavenger
Metal scavenger: Au, Cd, Co, Cu, Fe, Pd, Pt, Rh, Ru, V, Zn, Pb
Manufacturer: Alfa Aesar™ 4621806
Quantity 5g
 View more versions of this product
Catalog No. AA4621806
  N/A
Add to Cart
Related Products
Product Content Feedback